missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isocarbostyril, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC166460050

 View more versions of this product

Catalog No. AC166460050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Isocarbostyril
Glass bottle
211.0°C to 214.0°C
491-30-5
C9H7NO
1 2h-isoquinolinone
C1=CC=C2C(=C1)C=CNC2=O
145.16
CHEBI:18350
Powder or Crystalline Powder
Authentic
Brown to White
5g
97.5% min. (HPLC)
21,1
VDBNYAPERZTOOF-UHFFFAOYSA-N
2H-isoquinolin-1-one
10284
145.16
98%

Chemical Identifiers

491-30-5
145.16
1 2h-isoquinolinone
CHEBI:18350
C1=CC=C2C(=C1)C=CNC2=O
C9H7NO
VDBNYAPERZTOOF-UHFFFAOYSA-N
10284
2H-isoquinolin-1-one
Safety and Handling

Safety and Handling

EINECSNumber : 207-732-1

SDS Documents