Please login to your online account to display your discounted pricing

Isocarbostyril, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isocarbostyril
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H7NO
Formula Weight 145.16
Synonym 1-Hydroxyisoquinoline
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166460050

 
thermo fisher scientific
AC166460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166460010

 
thermo fisher scientific
AC166460010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Assay ≥97.5%
Formula Weight 145.16
Quantity 5g
Color White to pale brown
Melting Point 210° to 215°C
Chemical Name or Material Isocarbostyril
Assay Percent Range 98%
CAS 491-30-5
Synonym 1-Hydroxyisoquinoline
Molecular Formula C9H7NO
Infrared Spectrum Authentic
Packaging Glass bottle