Please login to your online account to display your discounted pricing

Isobutyrophenone 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  157111000

 View more versions of this product

Catalog No. AC157111000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 0.9880g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5162 to 1.5182 (20°C, 589nm)
Color Clear colorless to very slightly yellow
Quantity 100g
Chemical Name or Material Isobutyrophenone
Name Note 97%
CAS 611-70-1
Physical Form Clear colorless to very slightly yellow liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C10H12O
Linear Formula C6H5COCH(CH3)2
Formula Weight 148.20g/mol
MDL Number MFCD00008917
Beilstein 07, 316
EINECS Number 210-275-0
TSCA TSCA

Specifications

Chemical Name or Material Isobutyrophenone
Name Note 97%
CAS 611-70-1
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C10H12O
Linear Formula C6H5COCH(CH3)2
Formula Weight 148.20g/mol
MDL Number MFCD00008917