Please login to your online account to display your discounted pricing

Isobutyrophenone 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isobutyrophenone
Name Note 97%
CAS 611-70-1
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C10H12O
Linear Formula C6H5COCH(CH3)2
Formula Weight 148.20g/mol
MDL Number MFCD00008917
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC157111000

  SDS
fisher scientific acros
157111000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.9880g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5162 to 1.5182 (20°C, 589nm)
Color Clear colorless to very slightly yellow
Quantity 100g
Chemical Name or Material Isobutyrophenone
Name Note 97%
CAS 611-70-1
Physical Form Clear colorless to very slightly yellow liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C10H12O
Linear Formula C6H5COCH(CH3)2
Formula Weight 148.20g/mol
MDL Number MFCD00008917
Beilstein 07, 316
EINECS Number 210-275-0
TSCA TSCA