Please login to your online account to display your discounted pricing

Isobutyronitrile 99%, ACROS Organics™

$64.67 - $64.67

Chemical Identifiers

Chemical Name or Material Isobutyronitrile
Name Note 99%
CAS 78-82-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H7N
Linear Formula (CH3)2CHCN
Formula Weight 69.11
MDL Number MFCD00001873
Synonym 2-Methylpropionitrile
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC122530010

  SDS
thermo fisher scientific
122530010
Glass bottle 1L Each for $64.67
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 69.11
Quantity 1L
Color Clear colorless to light yellow
EINECS Number 201-147-5
TSCA TSCA
Density 0.76g/mL
RTECS Number TZ4900000
Beilstein 02, 294
Merck Index 15, 5201
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Isobutyronitrile
Name Note 99%
Assay Percent Range 99%
Molecular Formula C4H7N
Linear Formula (CH3)2CHCN
MDL Number MFCD00001873
CAS 78-82-0
Synonym 2-Methylpropionitrile
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.3720 to 1.3740 (20°C, 589nm)
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
Highly flammable liquid and vapor.