missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyraldehyde, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC122510100

 View more versions of this product

Catalog No. AC122510100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Isobutyraldehyde
−24°C
Plastic drum
0.79
63.0°C
-65.0°C
78-84-2
C4H8O
MFCD00006980
15, 5199
Solubility in water: 75g/L (20°C). Other solubilities: miscible with ethanol, ether, carbon disulfide,, acetone, benzene, toluene and chloroform
CC(C)C=O
72.11
CHEBI:48943
Liquid
0.7900g/mL
Authentic
1.3710 to 1.3740
0.45 mPa.s (20°C)
Undesignated
10L
99% min. (GC)
(CH3)2CHCHO
01, 671
2-methyl-1-propanal
AMIMRNSIRUDHCM-UHFFFAOYSA-N
2-methylpropanal
6561
72.11
99+%

Chemical Identifiers

78-84-2
72.11
AMIMRNSIRUDHCM-UHFFFAOYSA-N
6561
2-methylpropanal
C4H8O
MFCD00006980
2-methyl-1-propanal
CHEBI:48943
CC(C)C=O
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond conta

GHS Signal Word: Danger

EINECSNumber : 201-149-6

SDS Documents