Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyltriphenylphosphonium bromide, 98+%

$28.32 - $253.14

Chemical Identifiers

Chemical Name or Material Isobutyltriphenylphosphonium bromide
CAS 22884-29-3
Molecular Formula [(CH3)2CHCH2](C6H5)3PBr
Formula Weight 399.32
MDL Number MFCD00031593
Beilstein 4171228
Synonym (2-Methylpropyl)triphenylphosphonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1788106

  SDS
alfa aesar™
A1788106
5g Each for $28.32

AAA1788114

  SDS
alfa aesar™
A1788114
25g Each for $74.28

AAA1788122

  SDS
alfa aesar™
A1788122
100g Each for $253.14
Description & Specifications

Specifications

Formula Weight 399.32
Chemical Name or Material Isobutyltriphenylphosphonium bromide
Assay ≥98%
Molecular Formula [(CH3)2CHCH2](C6H5)3PBr
MDL Number MFCD00031593
CAS 22884-29-3
Beilstein 4171228
Synonym (2-Methylpropyl)triphenylphosphonium bromide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 198° to 202°C
Quantity 5g

Applications
It is employed as intermediate for pharmaceutical.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.