missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyltriethoxysilane, 95%

Manufacturer:  Alfa Aesar™ H5946114

 View more versions of this product

Catalog No. AAH5946114



Description

Description

Applications
It is used in a wide range of applications including silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent, silane cross linking agent, silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent, silane reinforcer, silylating agent, silane reducing agent, silyl building blocks and synthons.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Specifications

Specifications

Isobutyltriethoxysilane
C10H24O3Si
MFCD00069084
triethoxy isobutyl silane, isobutyltriethoxysilane, triethoxy 2-methylpropyl silane, triethoxyisobutylsilane, unii-0yz0g6z2xh, silane, triethoxy 2-methylpropyl, 0yz0g6z2xh, dynasylan ibteo, 2-methylpropyl triethoxysilane, iso-butyltriethoxysilane
CCO[Si](CC(C)C)(OCC)OCC
220.384
220.38
95%
1.4505
190°C to 191°C
17980-47-1
(CH3)2CHCH2Si(OC2H5)3
1750049
ALVYUZIFSCKIFP-UHFFFAOYSA-N
triethoxy(2-methylpropyl)silane
87391
Liquid
0.880g/mL
165°C (400mmHg)
25g

Chemical Identifiers

17980-47-1
220.384
ALVYUZIFSCKIFP-UHFFFAOYSA-N
87391
CCO[Si](CC(C)C)(OCC)OCC
C10H24O3Si
MFCD00069084
triethoxy isobutyl silane, isobutyltriethoxysilane, triethoxy 2-methylpropyl silane, triethoxyisobutylsilane, unii-0yz0g6z2xh, silane, triethoxy 2-methylpropyl, 0yz0g6z2xh, dynasylan ibteo, 2-methylpropyl triethoxysilane, iso-butyltriethoxysilane
triethoxy(2-methylpropyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.

GHS P Statement
P210-P280-P362-P321-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents