missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isobutyltrichlorosilane 98.0+%, TCI America™
$136.03 - $136.03
Chemical Identifiers
CAS | 18169-57-8 |
---|---|
Molecular Formula | C4H9Cl3Si |
Molecular Weight (g/mol) | 191.551 |
MDL Number | MFCD00053185 |
InChI Key | GBXOGFTVYQSOID-UHFFFAOYSA-N |
Synonym | isobutyltrichlorosilane, trichloroisobutylsilane, silane, trichloro 2-methylpropyl, trichloro isobutyl silane, trichloro 2-methylpropyl silane, i-butyltrichlorosilane, silane,trichloro 2-methylpropyl, acmc-209ei4, trichloro isobutyl silane #, gbxogftvyqsoid-uhfffaoysa |
PubChem CID | 87489 |
IUPAC Name | trichloro(2-methylpropyl)silane |
SMILES | CC(C)C[Si](Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I045225G
|
TCI America
I045225G |
25 g |
Each for $136.03
|
|
|||||
Chemical Identifiers
18169-57-8 | |
191.551 | |
GBXOGFTVYQSOID-UHFFFAOYSA-N | |
87489 | |
CC(C)C[Si](Cl)(Cl)Cl |
C4H9Cl3Si | |
MFCD00053185 | |
isobutyltrichlorosilane, trichloroisobutylsilane, silane, trichloro 2-methylpropyl, trichloro isobutyl silane, trichloro 2-methylpropyl silane, i-butyltrichlorosilane, silane,trichloro 2-methylpropyl, acmc-209ei4, trichloro isobutyl silane #, gbxogftvyqsoid-uhfffaoysa | |
trichloro(2-methylpropyl)silane |
Specifications
18169-57-8 | |
140°C | |
MFCD00053185 | |
1747 | |
GBXOGFTVYQSOID-UHFFFAOYSA-N | |
trichloro(2-methylpropyl)silane | |
87489 | |
≥98.0% (GC) | |
Isobutyltrichlorosilane |
Colorless | |
C4H9Cl3Si | |
25 g | |
isobutyltrichlorosilane, trichloroisobutylsilane, silane, trichloro 2-methylpropyl, trichloro isobutyl silane, trichloro 2-methylpropyl silane, i-butyltrichlorosilane, silane,trichloro 2-methylpropyl, acmc-209ei4, trichloro isobutyl silane #, gbxogftvyqsoid-uhfffaoysa | |
CC(C)C[Si](Cl)(Cl)Cl | |
191.551 | |
191.55 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-2041
TSCA : Yes