Please login to your online account to display your discounted pricing

Isobutyltrichlorosilane, 98%, ACROS Organics™


Quantity 5mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  338060050

 View more versions of this product

Catalog No. AC338060050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.56
Physical Form Liquid
Assay ≥97.5%
Quantity 5mL
Color Clear colorless to slightly yellow
Density 1.15g/mL
EINECS Number 242-053-4
TSCA TSCA
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Isobutyltrichlorosilane
CAS 18169-57-8
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 98%
MDL Number MFCD00053185
Linear Formula (Cl3)SiCHCH(CH3)
Name Note 98%
Molecular Formula C4H9Cl3Si

Specifications

Chemical Name or Material Isobutyltrichlorosilane
Name Note 98%
CAS 18169-57-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H9Cl3Si
Linear Formula (Cl3)SiCHCH(CH3)
Formula Weight 191.56
MDL Number MFCD00053185
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.

SDS & Safety Documents