Please login to your online account to display your discounted pricing

Isobutyllithium, 1.6M solution in heptane, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC377591000

 View more versions of this product

Catalog No. AC377591000

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
May cause drowsiness or dizziness. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Catches fire spontaneously if exposed to air. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment.
Health Hazard 1 Danger
Formula Weight 64.06
Physical Form Liquid
Quantity 100mL
Color Yellow
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.7g/mL
Assay 15 to 17% (as iso-BuLi)
CAS 64742-49-0
Chemical Name or Material Isobutyllithium
Molecular Formula C4H9Li
CAS 920-36-5
Linear Formula (CH3)2CHCH2Li
Additional Information Product may develop some turbidity or precipitate
Name Note 1.6M Solution in Heptane
MDL Number MFCD01863085

Specifications

Chemical Name or Material Isobutyllithium
Name Note 1.6M Solution in Heptane
CAS 920-36-5
CAS 64742-49-0
Assay 15 to 17% (as iso-BuLi)
Molecular Formula C4H9Li
Linear Formula (CH3)2CHCH2Li
Formula Weight 64.06
MDL Number MFCD01863085
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment.

GHS H Statement
May cause drowsiness or dizziness. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Catches fire spontaneously if exposed to air. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

SDS & Safety Documents