Please login to your online account to display your discounted pricing

Isobutyllithium, 1.6M solution in heptane, AcroSeal™, ACROS Organics™

$598.42 - $598.42

Chemical Identifiers

Chemical Name or Material Isobutyllithium
Name Note 1.6M Solution in Heptane
CAS 920-36-5
CAS 64742-49-0
Assay 15 to 17% (as iso-BuLi)
Molecular Formula C4H9Li
Linear Formula (CH3)2CHCH2Li
Formula Weight 64.06
MDL Number MFCD01863085
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377591000

  SDS
thermo fisher scientific
AC377591000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $598.42
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 64.06
Quantity 100mL
Color Yellow
Packaging AcroSeal™ Glass Bottle in Tin Can
Assay 15 to 17% (as iso-BuLi)
Density 0.7g/mL
CAS 64742-49-0
Chemical Name or Material Isobutyllithium
Name Note 1.6M Solution in Heptane
Molecular Formula C4H9Li
Linear Formula (CH3)2CHCH2Li
MDL Number MFCD01863085
CAS 920-36-5
Health Hazard 1 GHS Signal Word: Danger
Additional Information Product may develop some turbidity or precipitate
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Catches fire spontaneously if exposed to air.
Highly flammable liquid and vapor.
In
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not allow contact with air.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Catches fire spontaneously if exposed to air.
Highly flammable liquid and vapor.
In