Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutylhydrazine hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material Isobutylhydrazine hydrochloride
CAS 237064-47-0
Linear Formula (CH3)2CHCH2NHNH2·HCl
Formula Weight 124.61
MDL Number MFCD06245510
Synonym 2-Methylpropylhydrazine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6428703

  SDS
alfa aesar™
H6428703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6428706

  SDS
alfa aesar™
H6428706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64287MD

  SDS
alfa aesar™
H64287MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.61
Chemical Name or Material Isobutylhydrazine hydrochloride
Physical Form Solid
Assay 97%
Linear Formula (CH3)2CHCH2NHNH2·HCl
MDL Number MFCD06245510
CAS 237064-47-0
Synonym 2-Methylpropylhydrazine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
It finds its application in the discovery and characterization of mapk-activated protein kinase-2 prevention of activation inhibitors.

Solubility
Slightly soluble in water. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.