missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutylene oxide, 99+%

Manufacturer:  Alfa Aesar™ L0977314

 View more versions of this product

Catalog No. AAL0977314



Description

Description

Applications
Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.

Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Isobutylene oxide
Ether-like
25g
C4H8O
UN2924
isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
CC1(CO1)C
72.107
72.11
−32°C (−25°F)
51°C to 52°C
558-30-5
MFCD00066354
102408
GELKGHVAFRCJNA-UHFFFAOYSA-N
2,2-dimethyloxirane
11208
≥99%

Chemical Identifiers

558-30-5
72.107
GELKGHVAFRCJNA-UHFFFAOYSA-N
11208
CC1(CO1)C
C4H8O
MFCD00066354
isobutylene oxide, 1,2-epoxy-2-methylpropane, isobutene oxide, isobutylene epoxide, isobutyleneoxide, oxirane, 2,2-dimethyl, 1,2-epoxyisobutane, 1,1-dimethylethylene oxide, 1,2-isobutylene oxide, 2-methyl-1-propene oxide
2,2-dimethyloxirane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

RTECSNumber : TZ3580000

TSCA : TSCA

Documents