missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutylboronic Acid (contains varying amounts of Anhydride), TCI America™
$137.94 - $137.94
Chemical Identifiers
| CAS | 84110-40-7 |
|---|---|
| Molecular Formula | C4H11BO2 |
| Molecular Weight (g/mol) | 101.94 |
| MDL Number | MFCD00134156 |
| InChI Key | ZAZPDOYUCVFPOI-UHFFFAOYSA-N |
| PubChem CID | 2734395 |
Specifications
| 84110-40-7 | |
| White-Yellow | |
| MFCD00134156 | |
| isobutylboronic acid, isobutaneboronic acid, 2-methylpropyl boronic acid, 1-isobutaneboronic acid, 2-methylpropaneboronic acid, 2-methylprop-1-ylboronic acid, unii-khx76h7jhz, khx76h7jhz, 2-methyl-1-propylboronic acid, 2-methyl-1-propaneboronic acid | |
| CC(C)CB(O)O | |
| 101.94 | |
| 101.94 | |
| Isobutylboronic Acid (contains varying amounts of Anhydride) |
| 104°C | |
| C4H11BO2 | |
| 5 g | |
| ZAZPDOYUCVFPOI-UHFFFAOYSA-N | |
| (2-methylpropyl)boronic acid | |
| 2734395 | |
| Crystalline Powder |
Safety and Handling
TSCA : No