missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutylamine, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC122502500

 View more versions of this product

Catalog No. AC122502500



Description

Description

Specifications

Specifications

Isobutylamine
−9°C
Glass bottle
0.73
64.0°C to 71.0°C
-85.0°C
78-81-9
(CH3)2CHCH2NH2
04, 163
1-amino-2-methylpropane, 1-propanamine, 2-methyl, 2-methylpropanamine, 2-methylpropylamine, 3-methyl-2-propylamine, i-butylamine, iso-butylamine, isobutylamine, monoisobutylamine, valamine
KDSNLYIMUZNERS-UHFFFAOYSA-N
2-methylpropan-1-amine
6558
73.14
≥98.5%
0.7300g/mL
Authentic
1.397
0.6 mPa.s (20°C)
Colorless to Yellow
250mL
C4H11N
MFCD00008146
15, 5177
Solubility in water: miscible. Other solubilities: soluble in alcohol and ether
CC(C)CN
73.14
CHEBI:15997
Liquid

Chemical Identifiers

78-81-9
73.14
KDSNLYIMUZNERS-UHFFFAOYSA-N
6558
2-methylpropan-1-amine
C4H11N
MFCD00008146
1-amino-2-methylpropane, 1-propanamine, 2-methyl, 2-methylpropanamine, 2-methylpropylamine, 3-methyl-2-propylamine, i-butylamine, iso-butylamine, isobutylamine, monoisobutylamine, valamine
CHEBI:15997
CC(C)CN
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 201-145-4

SDS Documents