Learn More
Isobutylamine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 122500010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isobutylamine | |
-85.0°C | |
0.7300g/mL | |
−9°C | |
Glass bottle | |
1.397 | |
MFCD00008146 | |
04, 163 | |
15, 5177 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol and ether | |
CC(C)CN | |
73.14 | |
0.6 mPa.s (20°C) | |
73.14 | |
Liquid |
78-81-9 | |
Colorless to Yellow | |
64.0°C to 71.0°C | |
Authentic | |
C4H11N | |
(CH3)2CHCH2NH2 | |
1 L | |
0.73 | |
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine | |
KDSNLYIMUZNERS-UHFFFAOYSA-N | |
2-methylpropan-1-amine | |
6558 | |
CHEBI:15997 | |
≥98.5% |
Chemical Identifiers
78-81-9 | |
73.14 | |
KDSNLYIMUZNERS-UHFFFAOYSA-N | |
6558 | |
2-methylpropan-1-amine |
C4H11N | |
MFCD00008146 | |
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine | |
CHEBI:15997 | |
CC(C)CN |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 201-145-4
RUO – Research Use Only