missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutylamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A11017AK

 View more versions of this product

Catalog No. AAA11017AK



Description

Description

Isobutylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutylamine is used as pharmaceutical intermediate.

Solubility
Soluble in water (1000 mg/ml at 25°C), ether, alcohol, acetone and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications

Specifications

Isobutylamine
°C to 85°C
67°C to 69°C
99%
1.397
250 mL
385626
14,5132
Soluble in water (1000mg/ml at 25°C),ether,alcohol,acetone and benzene.
CC(C)CN
73.139
CHEBI:15997
99%
78-81-9
0.736 g/mL
-9°C (15°F)
C4H11N
MFCD00008146
UN1214
Air Sensitive
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
KDSNLYIMUZNERS-UHFFFAOYSA-N
2-methylpropan-1-amine
6558
73.14
Liquid

Chemical Identifiers

78-81-9
73.139
KDSNLYIMUZNERS-UHFFFAOYSA-N
6558
2-methylpropan-1-amine
C4H11N
MFCD00008146
isobutylamine, 1-amino-2-methylpropane, 2-methylpropylamine, monoisobutylamine, 1-propanamine, 2-methyl, valamine, iso-butylamine, i-butylamine, 2-methylpropanamine, 3-methyl-2-propylamine
CHEBI:15997
CC(C)CN
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H314-H318

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H301-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYLAMINE

EINECSNumber : 201-145-4

RTECSNumber : NP9900000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only