Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyl propionate, 98%

Chemical Identifiers

Chemical Name or Material Isobutyl propionate
CAS 540-42-1
Linear Formula CH3CH2CO2CH2CH(CH3)2
Formula Weight 130.18
MDL Number MFCD00009307
UN Number UN2394
Synonym Propionic acid isobutyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH59612AC

 
alfa aesar™
H59612AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH59612AE

 
alfa aesar™
H59612AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.18
Chemical Name or Material Isobutyl propionate
Density 0.869g/mL
Physical Form Liquid
Assay 98%
Linear Formula CH3CH2CO2CH2CH(CH3)2
MDL Number MFCD00009307
CAS 540-42-1
UN Number UN2394
Synonym Propionic acid isobutyl ester
Health Hazard 1 Warning
Refractive Index 1.3970
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Colorless
Melting Point −71°C
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ISOBUTYL PROPIONATE
RTECS Number UF4930000
Quantity 25mL
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.