Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyl p-toluenesulfonate, 97%

$167.26 - $167.26

Chemical Identifiers

Chemical Name or Material Isobutyl p-toluenesulfonate
CAS 4873-56-7
Molecular Formula C11H16O3S
Formula Weight 228.28
MDL Number MFCD00026478
Synonym p-Toluenesulfonic acid isobutyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2008014

  SDS
alfa aesar™
B2008014
25g Each for $167.26

AAB2008006

  SDS
alfa aesar™
B2008006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Isobutyl p-toluenesulfonate
Quantity 25g
CAS 4873-56-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C11H16O3S
Formula Weight 228.28
MDL Number MFCD00026478
Synonym p-Toluenesulfonic acid isobutyl ester
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.