Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyl nitrite, 95%, stab. with potassium carbonate

Chemical Identifiers

Chemical Name or Material Isobutyl nitrite
CAS 542-56-3
Name Note stabilized with potassium carbonate
Molecular Formula C4H9NO2
Linear Formula (CH3)2CHCH2ONO
Formula Weight 103.12
MDL Number MFCD00002056
UN Number UN2351
Beilstein 1699518
Merck Index 14,5149
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL05259AC

  SDS
alfa aesar™
L05259AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL05259AE

  SDS
alfa aesar™
L05259AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL05259AP

  SDS
alfa aesar™
L05259AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 103.12
Chemical Name or Material Isobutyl nitrite
CAS 542-56-3
MDL Number MFCD00002056
Molecular Formula C4H9NO2
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL NITRITES
TSCA TSCA
Assay 95%
Quantity 25mL
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H350-H341-H302-H332
Highly flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
UN Number UN2351
Recommended Storage 2° to 8°C
Synonym Nitrous acid isobutyl ester
Linear Formula (CH3)2CHCH2ONO
Name Note stabilized with potassium carbonate
Boiling Point 66° to 67°C
Merck Index 14,5149
Flash Point −21°C (−5°F)
Beilstein 1699518
RTECS Number RA0805000
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H350-H341-H302-H332
Highly flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.

2° to 8°C