missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl Myristate, TCI America™

Manufacturer:  TCI America M0555500ML

 View more versions of this product

Catalog No. M0555500ML



Specifications

Specifications

Isobutyl Myristate
500mL
C18H36O2
Myristic Acid Isobutyl Ester
CCCCCCCCCCCCCC(=O)OCC(C)C
284.484
284.48
≥97.0% (GC)
Colorless
25263-97-2
MFCD00059286
SKVCWXRLKHBEKW-UHFFFAOYSA-N
2-methylpropyl tetradecanoate
91361
Clear Liquid at 20°C

Chemical Identifiers

25263-97-2
284.484
SKVCWXRLKHBEKW-UHFFFAOYSA-N
91361
CCCCCCCCCCCCCC(=O)OCC(C)C
C18H36O2
MFCD00059286
Myristic Acid Isobutyl Ester
2-methylpropyl tetradecanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : No

Documents