missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl Myristate 97.0+%, TCI America™

$209.20 - $1,764.55

Chemical Identifiers

CAS 25263-97-2
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.484
MDL Number MFCD00059286
InChI Key SKVCWXRLKHBEKW-UHFFFAOYSA-N
Synonym Myristic Acid Isobutyl Ester
PubChem CID 91361
IUPAC Name 2-methylpropyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M055525ML
View Documents
Tci America
M055525ML
25mL Each for $209.20
 
M0555500ML
View Documents
Tci America
M0555500ML
500mL Each for $1,764.55
 
Specifications

Chemical Identifiers

25263-97-2
284.484
SKVCWXRLKHBEKW-UHFFFAOYSA-N
91361
CCCCCCCCCCCCCC(=O)OCC(C)C
C18H36O2
MFCD00059286
Myristic Acid Isobutyl Ester
2-methylpropyl tetradecanoate

Specifications

Colorless
25263-97-2
MFCD00059286
SKVCWXRLKHBEKW-UHFFFAOYSA-N
2-methylpropyl tetradecanoate
91361
Clear Liquid at 20°C
Isobutyl Myristate
25mL
C18H36O2
Myristic Acid Isobutyl Ester
CCCCCCCCCCCCCC(=O)OCC(C)C
284.484
284.48
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : No