missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl Methanesulfonate, TCI America™

$64.17 - $192.50

Chemical Identifiers

CAS 16156-53-9
Molecular Formula C5H12O3S
Molecular Weight (g/mol) 152.208
MDL Number MFCD07363982
InChI Key PZMKMCFRTKGVOL-UHFFFAOYSA-N
Synonym Methanesulfonic Acid Isobutyl Ester
PubChem CID 119240
IUPAC Name 2-methylpropyl methanesulfonate
SMILES CC(C)COS(=O)(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I10625G View Documents TCI America
I10625G
5g Each for $192.50
 
I10621G View Documents TCI America
I10621G
1g Each for $64.17
 
Specifications

Chemical Identifiers

16156-53-9
152.208
PZMKMCFRTKGVOL-UHFFFAOYSA-N
119240
CC(C)COS(=O)(=O)C
C5H12O3S
MFCD07363982
Methanesulfonic Acid Isobutyl Ester
2-methylpropyl methanesulfonate

Specifications

53°C
5g
C5H12O3S
Methanesulfonic Acid Isobutyl Ester
CC(C)COS(=O)(=O)C
152.208
152.21
≥98.0% (GC)
Yellow
16156-53-9
MFCD07363982
PZMKMCFRTKGVOL-UHFFFAOYSA-N
2-methylpropyl methanesulfonate
119240
Clear Liquid at 20°C
Isobutyl Methanesulfonate
Safety and Handling

Safety and Handling

RTECSNumber : PB2662500

TSCA : No