missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isobutyl Methacrylate Polymer, TCI America™
Supplier: TCI America M0086500G
Specifications
White | |
Crystal-Powder at 20°C | |
C8H14O2 | |
Methacrylic Acid Isobutyl Ester Polymer |
Isobutyl Methacrylate Polymer | |
9011-15-8 | |
MFCD00008931 | |
500 g |
Chemical Identifiers
9011-15-8 | |
C8H14O2 | |
MFCD00008931 | |
Methacrylic Acid Isobutyl Ester Polymer |
Safety and Handling
EINECSNumber : (6)-0524
TSCA : Yes