Please login to your online account to display your discounted pricing

Isobutyl methacrylate, 99%, ACROS Organics™


Quantity 2.5L
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC412680025

 View more versions of this product

Catalog No. AC412680025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 142.2
Quantity 2.5L
Color Clear colorless
Density 0.8800g/mL
Molecular Formula C8H14O2
Linear Formula CH2=C(CH3)COOCH2CH(CH3)2
CAS 97-86-9
Acidity 0.005% max. (as methacrylic acid)
Water 0.1% max.
Chemical Name or Material Isobutyl methacrylate
Name Note Stabilized
Assay Percent Range 99%
MDL Number MFCD00008931
Packaging Plastic bottle
Health Hazard 1 GHS Signal Word: Warning
Refractive Index 1.4190 to 1.4220 (20°C, 589nm)
Stabilizer 15 to 20ppm MEHQ
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Isobutyl methacrylate
Name Note Stabilized
CAS 97-86-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H14O2
Linear Formula CH2=C(CH3)COOCH2CH(CH3)2
Formula Weight 142.2
MDL Number MFCD00008931
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents