Learn More
Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 041948AK
Description
Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isobutyl methacrylate | |
97-86-9 | |
155°C | |
C8H14O2 | |
250 mL | |
1747595 | |
Soluble in water at 20°C,2g/L | |
CC(C)COC(=O)C(C)=C | |
142.20 | |
142.2 | |
Liquid |
Stabilized with 10ppm 4-methoxyphenol | |
0.889 g/mL | |
46°C (115°F) | |
MFCD00008931,MFCD00084435 | |
UN2283 | |
isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove | |
RUMACXVDVNRZJZ-UHFFFAOYSA-N | |
2-methylpropyl 2-methylprop-2-enoate | |
7352 | |
≥99.5% |
Chemical Identifiers
97-86-9 | |
142.20 | |
RUMACXVDVNRZJZ-UHFFFAOYSA-N | |
7352 | |
CC(C)COC(=O)C(C)=C |
C8H14O2 | |
MFCD00008931,MFCD00084435 | |
isobutyl methacrylate, 2-propenoic acid, 2-methyl-, 2-methylpropyl ester, methacrylic acid, isobutyl ester, isobutyl 2-methyl-2-propenoate, 2-methylpropyl methacrylate, isobutyl alpha-methacrylate, isobutyl alpha-methylacrylate, unii-v11534uyz0, ccris 4829, isobutylester kyseliny methakrylove | |
2-methylpropyl 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
H226-H315-H319-H317-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
H226-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL METHACRYLATE, STABILIZED
EINECSNumber : 202-613-0
RTECSNumber : OZ4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only