missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl Isovalerate, TCI America™

$51.95 - $323.89

Chemical Identifiers

CAS 589-59-3
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00048349
InChI Key KEBDNKNVCHQIJU-UHFFFAOYSA-N
Synonym Isovaleric Acid Isobutyl Ester
PubChem CID 11514
IUPAC Name 2-methylpropyl 3-methylbutanoate
SMILES CC(C)CC(=O)OCC(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0197500ML View Documents TCI America
I0197500ML
500mL Each for $323.89
 
I019725ML View Documents TCI America
I019725ML
25mL Each for $51.95
 
Specifications

Chemical Identifiers

589-59-3
158.241
KEBDNKNVCHQIJU-UHFFFAOYSA-N
11514
CC(C)CC(=O)OCC(C)C
C9H18O2
MFCD00048349
Isovaleric Acid Isobutyl Ester
2-methylpropyl 3-methylbutanoate

Specifications

168°C
500mL
C9H18O2
3272
KEBDNKNVCHQIJU-UHFFFAOYSA-N
2-methylpropyl 3-methylbutanoate
11514
Clear Liquid at 20°C
Isobutyl Isovalerate
Colorless
589-59-3
MFCD00048349
Isovaleric Acid Isobutyl Ester
CC(C)CC(=O)OCC(C)C
158.241
158.24
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0776

RTECSNumber : NY1506000

TSCA : Yes