Please login to your online account to display your discounted pricing

Isobutyl Isobutyrate 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412675000

 View more versions of this product

Catalog No. AC412675000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (as area percentage)
Physical Form Liquid
Formula Weight 144.21
Quantity 500g
Color Clear
Merck Index 15, 5190
TSCA TSCA
RTECS Number NQ5250000
EINECS Number 202-612-5
Density 0.856g/mL
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Infrared Spectrum Authentic
Refractive Index 1.3980 to 1.4000 (20°C, 589nm)
Packaging Glass bottle
Synonym IBIB
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 99%
MDL Number MFCD00008916
Linear Formula (CH3)2CHCOOCH2CH(CH3)2
Molecular Formula C8H16O2
CAS 97-85-8
Chemical Name or Material Isobutyl isobutyrate
Name Note 99%

Specifications

Chemical Name or Material Isobutyl isobutyrate
Name Note 99%
CAS 97-85-8
Assay ≥98.5% (as area percentage)
Assay Percent Range 99%
Molecular Formula C8H16O2
Linear Formula (CH3)2CHCOOCH2CH(CH3)2
Formula Weight 144.21
MDL Number MFCD00008916
Synonym IBIB
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents