missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl Isobutyrate 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 412670050

 View more versions of this product

Catalog No. AC412670050


 

Specifications Description & Specifications

Specifications

Isobutyl isobutyrate
99%
0.856g/mL
≥98.5% (as area percentage)
C8H16O2
144.21
15, 5190
Authentic
1.3980 to 1.4000 (20°C, 589nm)
5g
97-85-8
Liquid
99%
(CH3)2CHCOOCH2CH(CH3)2
MFCD00008916
IBIB
Glass bottle
Clear

Chemical Identifiers

97-85-8
C8H16O2
144.21
IBIB
≥98.5% (as area percentage)
99%
(CH3)2CHCOOCH2CH(CH3)2
MFCD00008916
Isobutyl isobutyrate, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-612-5

RTECSNumber : NQ5250000

TSCA : TSCA