Please login to your online account to display your discounted pricing

Isobutyl Isobutyrate 99%, ACROS Organics™

$40.45 - $64.57

Chemical Identifiers

Chemical Name or Material Isobutyl isobutyrate
Name Note 99%
CAS 97-85-8
Assay ≥98.5% (as area percentage)
Assay Percent Range 99%
Molecular Formula C8H16O2
Linear Formula (CH3)2CHCOOCH2CH(CH3)2
Formula Weight 144.21
MDL Number MFCD00008916
Synonym IBIB
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC412670050

  SDS
thermo fisher scientific
412670050
Glass bottle 5g Each for $40.45

AC412675000

  SDS
thermo fisher scientific
412675000
Glass bottle 500g Each for $64.57
Description & Specifications

Specifications

Assay ≥98.5% (as area percentage)
Formula Weight 144.21
Physical Form Liquid
Quantity 5g
Color Clear
RTECS Number NQ5250000
TSCA TSCA
EINECS Number 202-612-5
Density 0.856g/mL
Merck Index 15, 5190
Infrared Spectrum Authentic
Refractive Index 1.3980 to 1.4000 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Synonym IBIB
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Linear Formula (CH3)2CHCOOCH2CH(CH3)2
MDL Number MFCD00008916
Health Hazard 1 GHS Signal Word: Warning
Assay Percent Range 99%
Chemical Name or Material Isobutyl isobutyrate
Name Note 99%
Molecular Formula C8H16O2
CAS 97-85-8
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.