Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyl dodecanoate, 99+%

$26.23 - $245.61

Chemical Identifiers

Chemical Name or Material Isobutyl dodecanoate
CAS 37811-72-6
Formula Weight 256.43
MDL Number MFCD00048424
Synonym Isobutyl laurate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42023AC

  SDS
alfa aesar™
42023AC
25mL Each for $26.23

AA42023AE

  SDS
alfa aesar™
42023AE
100mL Each for $61.64

AA42023AP

  SDS
alfa aesar™
42023AP
500mL Each for $245.61
Description & Specifications

Specifications

Formula Weight 256.43
Chemical Name or Material Isobutyl dodecanoate
Density 0.86g/mL
Physical Form Liquid
Assay ≥99%
MDL Number MFCD00048424
CAS 37811-72-6
Synonym Isobutyl laurate
Flash Point 150° (302°F)
Quantity 25mL