missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl chloroformate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1469222*

 View more versions of this product

Catalog No. AAA1469222



Description

Description

Applications
Isobutyl chloroformate is used as a peptide reagent. It is also used in the preparation of phenethyl-carbamic acid isobutyl ester by reaction with phenethylamine.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, acids, bases, alcohols and amines. Moisture sensitive.
Specifications

Specifications

Isobutyl chloroformate
128°C to 129°C
100g
98%
ClCO2CH2CH(CH3)2
UN2742
14,5139
YOETUEMZNOLGDB-UHFFFAOYSA-N
2-methylpropyl carbonochloridate
62365
Liquid
30°C (86°F)
-80°C
543-27-1
C5H9ClO2
MFCD00000642
956590
isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
CC(C)COC(=O)Cl
136.575
136.58
98%

Chemical Identifiers

543-27-1
136.575
YOETUEMZNOLGDB-UHFFFAOYSA-N
62365
CC(C)COC(=O)Cl
C5H9ClO2
MFCD00000642
isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob
2-methylpropyl carbonochloridate
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROFORMATES, TOXIC, CORROSIVE, FLAMMABLE, N.O.S.

TSCA : TSCA

storageNote1 : Moisture sensitive

Documents