Learn More
Isobutyl chloroformate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 176185000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentSpecifications
Isobutyl chloroformate | |
543-27-1 | |
128.7°C | |
Authentic | |
Glass bottle | |
1.4060 to 1.4080 | |
MFCD00000642 | |
03, 12 | |
1.0475 | |
isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob | |
YOETUEMZNOLGDB-UHFFFAOYSA-N | |
2-methylpropyl carbonochloridate | |
62365 | |
98% |
98% | |
1.0475g/mL | |
30°C | |
1% max. diisobutyl carbonate (GC) | |
C5H9ClO2 | |
ClCO2CH2CH(CH3)2 | |
500 g | |
06,307 | |
15, 5184 | |
Solubility in water: decomposes. Other solubilities: decomposes in alcohol,miscible with benzene,chloroform,acetone,toluene | |
CC(C)COC(=O)Cl | |
136.58 | |
136.58 | |
Liquid |
Chemical Identifiers
543-27-1 | |
136.58 | |
YOETUEMZNOLGDB-UHFFFAOYSA-N | |
62365 | |
CC(C)COC(=O)Cl |
C5H9ClO2 | |
MFCD00000642 | |
isobutyl chloroformate, 2-methylpropyl chloroformate, chloroformic acid isobutyl ester, isobutyl chlorocarbonate, carbonochloridic acid, 2-methylpropyl ester, isobutylchloroformate, i-butyl chloroformate, chlorocarbonic acid isobutyl ester, isobutyl carbonochloridate, unii-6s785tc0ob | |
2-methylpropyl carbonochloridate |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 208-840-1
TSCA : TSCA
RUO – Research Use Only