Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyl benzoate, 99%

$56.81 - $197.03

Chemical Identifiers

Chemical Name or Material Isobutyl benzoate
CAS 120-50-3
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00048344
Beilstein 2045961
Synonym Benzoic acid isobutyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA101140B

 
alfa aesar™
A101140B
1000g Each for $197.03

AAA1011430

 
alfa aesar™
A1011430
250g Each for $56.81
Description & Specifications

Specifications

Formula Weight 178.23
Chemical Name or Material Isobutyl benzoate
TSCA TSCA
Assay 99%
Molecular Formula C11H14O2
MDL Number MFCD00048344
CAS 120-50-3
Beilstein 2045961
Synonym Benzoic acid isobutyl ester
Boiling Point 242°C
Flash Point 96°C (204°F)
RTECS Number DH4185000
Quantity 1000g