missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Isobutyl benzoate, 99%

Manufacturer:  Alfa Aesar™ A1011430

 View more versions of this product

Catalog No. AAA1011430



Specifications

Specifications

Isobutyl benzoate
242°C
120-50-3
MFCD00048344
isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate
CC(C)COC(=O)C1=CC=CC=C1
178.231
CHEBI:87500
99%
96°C (204°F)
250g
C11H14O2
2045961
KYZHGEFMXZOSJN-UHFFFAOYSA-N
2-methylpropyl benzoate
61048
178.23

Chemical Identifiers

120-50-3
178.231
KYZHGEFMXZOSJN-UHFFFAOYSA-N
61048
2-methylpropyl benzoate
C11H14O2
MFCD00048344
isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate
CHEBI:87500
CC(C)COC(=O)C1=CC=CC=C1
Safety and Handling

Safety and Handling

RTECSNumber : DH4185000

TSCA : TSCA

SDS Documents