Please login to your online account to display your discounted pricing

Isobutyl Alcohol (Certified ACS), Fisher Chemical

$297.00 - $740.00

Chemical Identifiers

Chemical Name or Material Isobutyl Alcohol
CAS 78-83-1
Purity Grade Certified ACS Grade
Assay ≥99.0%
Assay Percent Range ≥99.0 %
Molecular Formula C4H10O
Linear Formula (CH3)2CHCH2OH
Formula Weight 74.12
MDL Number MFCD00004740
Synonym 1-Hydroxymethylpropane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A397-1

  SDS
fisher scientific company
BPA3971
Glass Bottle 1L Each for $297.00
Case of 6 Each for $1,782.00

A397-4

  Accessories SDS
fisher scientific company
BPA3974
Glass Bottle 4L Each for $740.00
Case of 4 Each for $2,960.00
Description & Specifications

Specifications

Assay ≥99.0%
Purity Grade Certified ACS Grade
Formula Weight 74.12
Boiling Point 108°C
Melting Point −108°C
Quantity 1L
UN Number 1120
Flash Point 28°C
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 11.7mbar at 20°C
Physical Form Liquid
Density 0.800g/cm³
Molecular Formula C4H10O
Color Colorless
Residue after Evaporation 0.001% max.
Solubility in Water Pass Test
Cleanup Symbol Solvent
Assay Percent Range ≥99.0 %
Identification Pass Test
MDL Number MFCD00004740
Synonym 1-Hydroxymethylpropane
Packaging Glass Bottle
Additional Information Vapor Pressure: 11.7mbar at 20°C
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Irritating to respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Seek immediate medical attention/advice. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:3
Instability:0
Butyraldehyde 0.01% max.
2_Butanone 0.02% max.
Chemical Name or Material Isobutyl Alcohol
Fluorescence Background Pass Test
Linear Formula (CH3)2CHCH2OH
CAS 78-83-1
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Water 0.1% max.
Titratable Acid 0.0005mEq/g max.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. Irritating to respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Seek immediate medical attention/advice. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:3
Instability:0

DANGER!