Please login to your online account to display your discounted pricing

Isobutyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$122.84 - $270.67

Chemical Identifiers

Chemical Name or Material Isobutyl Alcohol
CAS 78-83-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3)2CHCH2OH
Formula Weight 74.12
Synonym 2-butanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003285

 
avantor performance
904401
Narrow Mouth Amber Glass Bottle 500mL Each for $122.84

02003284

 
avantor performance
904403
Narrow Mouth Amber Glass Bottle 4L Each for $270.67
Description & Specifications

Specifications

Formula Weight 74.12
Purity Grade Baker Analyzed™ ACS Reagent
CAS 78-83-1
Chemical Name or Material Isobutyl Alcohol
Molecular Formula (CH3)2CHCH2OH
Synonym 2-butanol
Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle
Flash Point 28.00°C
Specific Gravity 1L = 0.80kg