Learn More
Isobutyl acrylate, 99%, stab with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L12739AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isobutyl acrylate | |
106-63-8 | |
0.888 | |
32°C (89°F) | |
1.415 | |
MFCD00042865 | |
UN2527 | |
isobutyl acrylate, isobutyl propenoate, isobutyl 2-propenoate, 2-propenoic acid, 2-methylpropyl ester, acrylic acid, isobutyl ester, 2-methylpropyl acrylate, isobutylester kyseliny akrylove, propenoic acid, isobutyl ester, ccris 4828, hsdb 610 | |
CC(C)COC(=O)C=C | |
128.17 | |
128.17 |
stabilized with 100ppm 4-methoxyphenol | |
-61°C | |
139°C | |
C7H12O2 | |
CH2=CHCO2CH2CH(CH3)2 | |
500 mL | |
1749388 | |
CFVWNXQPGQOHRJ-UHFFFAOYSA-N | |
2-methylpropyl prop-2-enoate | |
7819 | |
99% |
Chemical Identifiers
106-63-8 | |
128.17 | |
CFVWNXQPGQOHRJ-UHFFFAOYSA-N | |
7819 | |
CC(C)COC(=O)C=C |
C7H12O2 | |
MFCD00042865 | |
isobutyl acrylate, isobutyl propenoate, isobutyl 2-propenoate, 2-propenoic acid, 2-methylpropyl ester, acrylic acid, isobutyl ester, 2-methylpropyl acrylate, isobutylester kyseliny akrylove, propenoic acid, isobutyl ester, ccris 4828, hsdb 610 | |
2-methylpropyl prop-2-enoate |
Safety and Handling
GHS H Statement
H226-H312-H332-H315-H317
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
P210-P233-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
H226-H312+H332-H315-H317-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL ACRYLATE, STABILIZED
EINECSNumber : 203-417-8
RTECSNumber : AT2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only