missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isobutyl acetate, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC259830250
Specifications
Isobutyl acetate | |
18°C | |
Metal Drum | |
0.873 | |
0.03% max. | |
Undesignated | |
25L | |
97.5% min. (GC) | |
CH3CO2CH2CH(CH3)2 | |
02, 131 | |
2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetate d'isobutyle, acetic acid isobutyl ester, acetic acid, 2-methylpropyl ester, acetic acid, isobutyl ester, beta-methylpropyl ethanoate, iso-butyl acetate, isobutyl acetate, isobutyl ethanoate | |
GJRQTCIYDGXPES-UHFFFAOYSA-N | |
2-methylpropyl acetate | |
8038 | |
116.16 | |
98% |
0.8730g/mL | |
Authentic | |
1.3890 to 1.3910 | |
0.7 mPa.s (20°C) | |
116.0°C to 117.5°C | |
-99.0°C | |
110-19-0 | |
C6H12O2 | |
MFCD00008932 | |
15, 5175 | |
Solubility in water: 7g/L (20°C). Other solubilities: freely soluble in alcohol,ether,propylene glycol,and other organic solvents | |
CC(C)COC(=O)C | |
116.16 | |
CHEBI:50569 | |
Liquid |
Chemical Identifiers
110-19-0 | |
116.16 | |
GJRQTCIYDGXPES-UHFFFAOYSA-N | |
8038 | |
2-methylpropyl acetate |
C6H12O2 | |
MFCD00008932 | |
2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetate d'isobutyle, acetic acid isobutyl ester, acetic acid, 2-methylpropyl ester, acetic acid, isobutyl ester, beta-methylpropyl ethanoate, iso-butyl acetate, isobutyl acetate, isobutyl ethanoate | |
CHEBI:50569 | |
CC(C)COC(=O)C |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May cause drowsiness or dizziness.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Take off immediately all con
GHS Signal Word: Danger
EINECSNumber : 203-745-1