Learn More
Isobutyl acetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 259830025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isobutyl acetate | |
-99.0°C | |
116.0°C to 117.5°C | |
Authentic | |
Glass bottle | |
1.3890 to 1.3910 | |
MFCD00008932 | |
02, 131 | |
15, 5175 | |
Solubility in water: 7g/L (20°C). Other solubilities: freely soluble in alcohol,ether,propylene glycol,and other organic solvents | |
CC(C)COC(C)=O | |
116.16 | |
0.7 mPa.s (20°C) | |
116.16 | |
Liquid |
110-19-0 | |
0.8730g/mL | |
18°C | |
97.5% min. (GC) | |
C6H12O2 | |
CH3CO2CH2CH(CH3)2 | |
2.5 L | |
0.873 | |
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate | |
GJRQTCIYDGXPES-UHFFFAOYSA-N | |
2-methylpropyl acetate | |
8038 | |
CHEBI:50569 | |
98% |
Chemical Identifiers
110-19-0 | |
116.16 | |
GJRQTCIYDGXPES-UHFFFAOYSA-N | |
8038 | |
2-methylpropyl acetate |
C6H12O2 | |
MFCD00008932 | |
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate | |
CHEBI:50569 | |
CC(C)COC(C)=O |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May cause drowsiness or dizziness.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Take off immediately all con
GHS Signal Word: Danger
EINECSNumber : 203-745-1
RUO – Research Use Only