missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutyl acetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A14892AK

 View more versions of this product

Catalog No. AAA14892AK



Description

Description

Isobutyl acetate is used as a solvent for lacquer, thinners, sealants, perfumes and nitrocellulose. Further, it is used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isobutyl acetate is used as a solvent for lacquer, thinners, sealants, perfumes and nitrocellulose. Further, it is used as a flavoring agent.

Solubility
Miscible with ethanol, ether, methanol and acetone. Slightly miscible with water and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Isobutyl acetate
-99°C
116°C to 117°C
98%
C6H12O2
MFCD00008932
UN1213
14,5130
Miscible with ethanol,ether,methanol and acetone. Slightly miscible with water and carbon tetrachloride.
CC(C)COC(C)=O
116.16
CHEBI:50569
98%
110-19-0
0.87
18°C (64°F)
Fruit-like
1.3895
250 mL
1741909
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
GJRQTCIYDGXPES-UHFFFAOYSA-N
2-methylpropyl acetate
8038
116.16
Liquid

Chemical Identifiers

110-19-0
116.16
GJRQTCIYDGXPES-UHFFFAOYSA-N
8038
2-methylpropyl acetate
C6H12O2
MFCD00008932
isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
CHEBI:50569
CC(C)COC(C)=O
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H225-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOBUTYL ACETATE

EINECSNumber : 203-745-1

RTECSNumber : AI4022500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only