missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isobutanol, Spectrophotometric Grade, >99%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.10%

Manufacturer:  Alfa Aesar 32433K7

 View more versions of this product

Catalog No. AA32433K7



Specifications

Specifications

Isobutanol
>99%
MFCD00004740
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
CC(C)CO
74.123
CHEBI:46645
Liquid
390°C (734°F)
4-40 hPa (3-30mm Hg) at 20°C
98°C to 99°C
-107°C
78-83-1
C4H10O
1262
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
2-methylpropan-1-ol
6560
74.12
Sprectrophotometric
Sweet
-12°C (-10.40°F)
Colorless
4L

Chemical Identifiers

78-83-1
74.123
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
6560
2-methylpropan-1-ol
C4H10O
MFCD00004740
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
CHEBI:46645
CC(C)CO
Safety and Handling

Safety and Handling

H318-H315-H226-H335-H336: Causes serious eye damage. Causes skin irritation. Flammable liquid and vapour. May cause respiratory irritation. May cause drowsiness or dizziness.

DOTInformation : Hazard Class: 3, Packing Group: III

EINECSNumber : 201-148-0

SDS Documents