Learn More
Isobutanol, HPLC Grade, 99+%, Thermo Scientific Chemicals
Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05%
Supplier: Thermo Scientific Chemicals 022908M1
Description
Isobutanol is a reagent used in organic reactions. It is employed in the synthesis of new fluorinating reagents. It is used as a mobile phase in High Performance Liquid Chromatography. It is also used in the lipase-catalyzed production of biodiesel as an energy source. It acts as a catalyst for liquid-phase esterification of acetic acid by ion-exchange resins. Dehydration of fermented isobutanol is useful for the production of renewable chemicals and fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Isobutanol | |
°C to 108°C | |
0.802 | |
28°C (82°F) | |
>99% | |
C4H10O | |
MFCD00004740 | |
1730878 | |
Miscible with alcohol and diethyl ether. Partially miscible with water. | |
CC(C)CO | |
74.123 | |
CHEBI:46645 | |
4-40 hPa (3-30mm Hg) at 20°C | |
Liquid |
78-83-1 | |
Colorless | |
107°C to 108°C | |
390°C (734°F) | |
Sweet | |
1.396 | |
4 x 1 L | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
2-methylpropan-1-ol | |
6560 | |
74.12 | |
HPLC |
Chemical Identifiers
78-83-1 | |
74.123 | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
6560 | |
2-methylpropan-1-ol |
C4H10O | |
MFCD00004740 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
CHEBI:46645 | |
CC(C)CO |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H318-H335-H336
DOTInformation : Hazard Class: 3, Packing Group: III
EINECSNumber : 201-148-0
RTECSNumber : NP9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only