Learn More
Isobornyl methacrylate, 85-90%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 292090010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Isobornyl methacrylate | |
7534-94-3 | |
245.0°C | |
Authentic | |
Glass bottle | |
1.4760 to 1.4780 | |
1kg | |
ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate | |
IAXXETNIOYFMLW-UHFFFAOYNA-N | |
[(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate | |
100 to 200ppm MEHQ | |
222.33 |
85-90% | |
0.9800g/mL | |
101°C | |
85% min. (GC) | |
C14H22O2 | |
MFCD00081070 | |
0.98 | |
Solubility in water: insoluble | |
CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C | |
222.33 | |
71311141 | |
85-90% |
Chemical Identifiers
7534-94-3 | |
222.33 | |
IAXXETNIOYFMLW-UHFFFAOYNA-N | |
71311141 | |
CC(=C)C(=O)OC1CC2CCC1(C)C2(C)C |
C14H22O2 | |
MFCD00081070 | |
ibma, isobornyl methacrylate, methacrylic acid isobornyl ester, iso-bornyl methacrylate, isobornyl methacrylate, technical grade, isobornyl methacrylate, stabilized with mehq, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, 2-yl methacrylate, 1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate | |
[(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 231-403-1
TSCA : TSCA
RUO â Research Use Only