missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isobornyl methacrylate, 85-90%, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 292092500
Specifications
Isobornyl methacrylate | |
101°C | |
Glass bottle | |
0.98 | |
245.0°C | |
-60.0°C | |
85-90% | |
85% min. (GC) | |
MFCD00081070 | |
Solubility in water: insoluble | |
CC(=C)C(=O)OC1CC2CCC1(C2(C)C)C | |
222.33 | |
222.33 | |
85-90% |
0.9800g/mL | |
Authentic | |
1.4760 to 1.4780 | |
100 to 200ppm MEHQ | |
Colorless to Yellow | |
250g | |
7534-94-3 | |
C14H22O2 | |
1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate, 2-yl methacrylate, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, ibma, iso-bornyl methacrylate, isobornyl methacrylate, isobornyl methacrylate, stabilized with mehq, isobornyl methacrylate, technical grade, methacrylic acid isobornyl ester | |
IAXXETNIOYFMLW-JENJKZFGSA-N | |
[(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate | |
71311141 | |
Liquid |
Chemical Identifiers
7534-94-3 | |
222.33 | |
IAXXETNIOYFMLW-JENJKZFGSA-N | |
71311141 | |
CC(=C)C(=O)OC1CC2CCC1(C2(C)C)C |
C14H22O2 | |
MFCD00081070 | |
1r,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl 2-methylprop-2-enoate, 2-yl methacrylate, exo-1,7,7-trimethylbicyclo 2.2.1 hept-, ibma, iso-bornyl methacrylate, isobornyl methacrylate, isobornyl methacrylate, stabilized with mehq, isobornyl methacrylate, technical grade, methacrylic acid isobornyl ester | |
[(1R,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 231-403-1
TSCA : TSCA