Learn More
Isobornyl acetate, 94%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L11748AP
Description
Isobornyl acetate is one of the most important chemicals used in the perfumery industry. It is used in toiletries and soaps as a flavoring agent and antiseptics. One of main applications is as an intermediate to produce camphor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsobornyl acetate is one of the most important chemicals used in the perfumery industry. It is used in toiletries and soaps as a flavoring agent and antiseptics. One of main applications is as an intermediate to produce camphor.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Isobornyl acetate | |
0.99 | |
89°C (192°F) | |
1.465 | |
500 mL | |
1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate | |
KGEKLUUHTZCSIP-JFGNBEQYSA-N | |
[(1S,3S,4S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate | |
6950273 | |
94% |
125-12-2 | |
220°C to 224°C | |
C12H20O2 | |
MFCD00867808,MFCD00135943 | |
3197572 | |
Not miscible or difficult to mix with water. | |
CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C | |
196.29 | |
196.29 |
Chemical Identifiers
125-12-2 | |
196.29 | |
KGEKLUUHTZCSIP-JFGNBEQYSA-N | |
6950273 | |
CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C |
C12H20O2 | |
MFCD00867808,MFCD00135943 | |
1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate | |
[(1S,3S,4S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 204-727-6
RTECSNumber : NP7350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only