Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)-Isoborneol, 95%

Chemical Identifiers

Chemical Name or Material (±)-Isoborneol
CAS 124-76-5
Molecular Formula C10H18O
Formula Weight 154.25
MDL Number MFCD00074821
UN Number UN1325
Beilstein 4126091
Merck Index 14,5128
Synonym exo-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2063222

  SDS
alfa aesar™
B2063222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.25
TSCA TSCA
Odor Strong
Chemical Name or Material (±)-Isoborneol
Health Hazard 1 Warning
RTECS Number NP7300000
Quantity 100g
Molecular Formula C10H18O
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
CAS 124-76-5
Synonym exo-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P305+P351+P338-P362
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
UN Number UN1325
Health Hazard 2 GHS H Statement
H228-H315-H319
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
Assay 95%
Beilstein 4126091
Melting Point ≈210°C (subl.)
Flash Point 74°C (165°F)
MDL Number MFCD00074821
Merck Index 14,5128

Applications
Used as a synthetic flavor, as a moth repellent, cold sore topical medication, muscle liniment, and steam-inhaled cough suppressant. It is also used in Used in perfumes.

Solubility
Insoluble in water. Soluble in dipropylene glycol, ethyl alcohol (1gm. in 2ml. 70% alcohol).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.