Please login to your online account to display your discounted pricing

Isoamyl propionate 98+%, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  385530500

 View more versions of this product

Catalog No. AC385530500

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 0.8690g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4050 to 1.4080 (20°C, 589nm)
Color Clear colorless
Quantity 50mL
Chemical Name or Material Isoamyl propionate
CAS 105-68-0
Physical Form Clear colorless liquid
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C8H16O2
Linear Formula CH3CH2C(O)O(CH2)2CH(CH3)2
Formula Weight 144.21g/mol
MDL Number MFCD00048611
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Isoamyl propionate
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C8H16O2
Linear Formula CH3CH2C(O)O(CH2)2CH(CH3)2
Formula Weight 144.21g/mol
MDL Number MFCD00048611
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents