Please login to your online account to display your discounted pricing

Isoamyl nitrite, 96%, stabilized, ACROS Organics™

$67.31 - $67.31

Chemical Identifiers

Chemical Name or Material Isoamyl nitrite
Name Note 90%
CAS 110-46-3
CAS 497-19-8
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C5H11NO2
Linear Formula (CH3)2CHCH2CH2ONO
Formula Weight 117.15
MDL Number MFCD00002057
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC412751000

  SDS
thermo fisher scientific
412751000
Plastic Bottle in Tin Can 100g Each for $67.31
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 117.15
Physical Form Liquid
Quantity 100g
Color Clear yellow
Packaging Plastic Bottle in Tin Can
TSCA TSCA
EINECS Number 203-770-8
RTECS Number NT0187500
Density 0.8700g/mL
Fieser 01,40; 04,270; 05,358; 06,307; 09,258
Merck Index 15, 5168
Beilstein 01, 402
Refractive Index 1.3840 to 1.3880 (20°C, 589nm)
CAS 497-19-8
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Health Hazard 1 Danger
Infrared Spectrum Authentic
Stabilizer 0.5% sodium carbonate
Synonym Isopentyl nitrite
Molecular Formula C5H11NO2
Chemical Name or Material Isoamyl nitrite
Name Note 90%
MDL Number MFCD00002057
CAS 110-46-3
Linear Formula (CH3)2CHCH2CH2ONO
Assay Percent Range 96%
Safety and Handling

Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.